The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256732.1 | NP_001243661.1 | 11 - 12 | 959 | 102 |
NM_001256733.1 | NP_001243662.1 | 10 - 11 | 875 | 102 | |
NM_001256734.1 | NP_001243663.1 | 10 - 11 | 869 | 102 | |
NM_001256735.1 | NP_001243664.1 | 10 - 11 | 845 | 102 | |
NM_001256736.1 | NP_001243665.1 | 10 - 11 | 845 | 102 | |
NM_012446.3 | NP_036578.2 | 11 - 12 | 935 | 102 | |
XM_011543297.2 | XP_011541599.1 | 11 - 12 | 874 | 102 | |
XM_017009304.1 | XP_016864793.1 | 11 - 12 | 777 | 102 | |
XM_017009305.1 | XP_016864794.1 | 12 - 13 | 866 | 102 | |
XM_017009306.1 | XP_016864795.1 | 11 - 12 | 850 | 102 | |
XM_017009307.1 | XP_016864796.1 | 10 - 11 | 663 | 102 | |
XM_017009308.1 | XP_016864797.1 | 12 - 13 | 1038 | 102 | |
XM_017009309.1 | XP_016864798.1 | 11 - 12 | 1102 | 102 | |
XM_017009310.1 | XP_016864799.1 | 11 - 12 | 1250 | 102 | |
GenBank mRNA | AF077048.1 | - | 11 - 12 | 799 | 102 |
AF161465.1 | - | 11 - 12 | 765 | 102 | |
AK074537.1 | - | 11 - 12 | 730 | 102 | |
AK293544.1 | - | 10 - 11 | 670 | 102 | |
AK297494.1 | - | 11 - 12 | 643 | 102 | |
AK299858.1 | - | 10 - 11 | 713 | 102 | |
AK312337.1 | - | 11 - 12 | 779 | 102 | |
AL080076.1 | - | 11 - 12 | 783 | 102 | |
BC017020.1 | - | 11 - 12 | 783 | 102 | |
CR936787.1 | - | 10 - 11 | 690 | 102 | |
DA202987.1 | - | 3 - 4 | 125 | 102 |