The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199263.1 | NP_001186192.1 | 10 - 11 | 1071 | 104 |
XM_006711368.3 | XP_006711431.1 | 11 - 12 | 1238 | 104 | |
XM_006711369.2 | XP_006711432.1 | 11 - 12 | 1235 | 104 | |
XM_017001433.1 | XP_016856922.1 | 10 - 11 | 1157 | 104 | |
XM_017001444.1 | XP_016856933.1 | 10 - 11 | 1319 | 104 | |
GenBank mRNA | AF151828.1 | - | 10 - 11 | 951 | 104 |
AF157320.1 | - | 10 - 11 | 1086 | 104 |