The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004564.2 | NP_004555.1 | 5 - 6 | 809 | 66 |
XM_005263063.3 | XP_005263120.1 | 5 - 6 | 771 | 66 | |
GenBank mRNA | AB019410.2 | - | 5 - 6 | 753 | 66 |
AF026851.1 | - | 5 - 6 | 780 | 66 | |
AF151033.1 | - | 5 - 6 | 777 | 66 | |
AK222887.1 | - | 5 - 6 | 771 | 66 | |
AK225241.1 | - | 5 - 6 | 771 | 66 | |
AK312957.1 | - | 5 - 6 | 771 | 66 | |
BC130348.1 | - | 5 - 6 | 819 | 66 | |
BC136547.1 | - | 5 - 6 | 819 | 66 | |
BX537555.1 | - | 3 - 4 | 5383 | 66 |