The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152751.2 | NP_689964.2 | 2 - 3 | 282 | 109 |
XM_011519397.2 | XP_011517699.1 | 2 - 3 | 282 | 109 | |
GenBank mRNA | AK097602.1 | - | 2 - 3 | 282 | 109 |