The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005245246.4 | XP_005245303.1 | 11 - 12 | 1230 | 144 |
XM_006711367.3 | XP_006711430.1 | 12 - 13 | 1314 | 144 | |
XM_017001429.1 | XP_016856918.1 | 12 - 13 | 1311 | 144 | |
XM_017001431.1 | XP_016856920.1 | 11 - 12 | 1233 | 144 | |
GenBank mRNA | AK225794.1 | - | 11 - 12 | 1158 | 219 |