The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020877.3 | NP_065928.2 | 17 - 18 | 4240 | 71 |
XM_011523663.1 | XP_011521965.1 | 19 - 20 | 3108 | 71 | |
XM_011523664.2 | XP_011521966.1 | 20 - 21 | 3257 | 71 | |
XM_011523666.1 | XP_011521968.1 | 19 - 20 | 3108 | 71 | |
XM_011523667.2 | XP_011521969.1 | 18 - 19 | 3011 | 71 | |
XM_011523668.1 | XP_011521970.1 | 9 - 10 | 1491 | 71 | |
XM_011523669.2 | XP_011521971.1 | 7 - 8 | 1175 | 71 | |
XM_011523670.2 | XP_011521972.1 | 19 - 20 | 3108 | 71 | |
XM_017024218.1 | XP_016879707.1 | 6 - 7 | 1192 | 71 | |
XM_017024219.1 | XP_016879708.1 | 19 - 20 | 3108 | 71 | |
GenBank mRNA | AB040936.2 | - | 17 - 18 | 4240 | 71 |