Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 57 - 58 10292 77
XM_011523663.1 XP_011521965.1 59 - 60 9160 77
XM_011523664.2 XP_011521966.1 60 - 61 9309 77
XM_011523666.1 XP_011521968.1 58 - 59 8971 77
XM_011523667.2 XP_011521969.1 58 - 59 9063 77
XM_011523668.1 XP_011521970.1 49 - 50 7543 77
XM_011523669.2 XP_011521971.1 47 - 48 7227 77
XM_011523670.2 XP_011521972.1 59 - 60 9160 77
XM_017024218.1 XP_016879707.1 46 - 47 7244 77
XM_017024219.1 XP_016879708.1 59 - 60 9160 77
GenBank mRNA AB040936.2 - 57 - 58 10292 77

View Full Product Details