The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303270.1 | NP_001290199.1 | 7 - 8 | 1058 | 98 |
NM_020877.3 | NP_065928.2 | 7 - 8 | 2432 | 98 | |
XM_011523663.1 | XP_011521965.1 | 7 - 8 | 1054 | 98 | |
XM_011523664.2 | XP_011521966.1 | 8 - 9 | 1203 | 98 | |
XM_011523666.1 | XP_011521968.1 | 7 - 8 | 1054 | 98 | |
XM_011523667.2 | XP_011521969.1 | 8 - 9 | 1203 | 98 | |
XM_011523670.2 | XP_011521972.1 | 7 - 8 | 1054 | 98 | |
XM_017024219.1 | XP_016879708.1 | 7 - 8 | 1054 | 98 | |
GenBank mRNA | AB040936.2 | - | 7 - 8 | 2432 | 98 |
AK292607.1 | - | 7 - 8 | 1054 | 98 | |
BC034225.1 | - | 7 - 8 | 1057 | 98 |