Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAK1
Entrez Gene ID
22848
Gene Name
AP2 associated kinase 1
Gene Aliases
-
Chromosome Location
Chr.2: 69457995 - 69643845 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014911.3 NP_055726.3 8 - 9 1368 99
GenBank mRNA AB028971.1 - 8 - 9 1247 99
AK056043.1 - 8 - 9 1175 99
BC002695.2 - 7 - 8 903 99
BC104842.1 - 7 - 8 887 99
BC111965.1 - 7 - 8 887 99
BC143710.1 - 7 - 8 887 99

View Full Product Details