The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018398.2 | NP_060868.2 | 16 - 17 | 1606 | 94 |
XM_005265318.2 | XP_005265375.1 | 15 - 16 | 1660 | 94 | |
XM_011533946.2 | XP_011532248.1 | 16 - 17 | 1678 | 94 | |
XM_011533947.2 | XP_011532249.1 | 16 - 17 | 1678 | 94 | |
XM_011533948.2 | XP_011532250.1 | 16 - 17 | 1678 | 94 | |
XM_011533949.2 | XP_011532251.1 | 16 - 17 | 1678 | 94 | |
XM_011533950.2 | XP_011532252.1 | 16 - 17 | 1678 | 94 | |
XM_011533951.2 | XP_011532253.1 | 16 - 17 | 1678 | 94 | |
XM_011533952.2 | XP_011532254.1 | 16 - 17 | 1678 | 94 | |
XM_011533953.2 | XP_011532255.1 | 15 - 16 | 1660 | 94 | |
XM_011533954.2 | XP_011532256.1 | 3 - 4 | 899 | 94 | |
XM_017006850.1 | XP_016862339.1 | 14 - 15 | 1588 | 94 | |
XM_017006851.1 | XP_016862340.1 | 13 - 14 | 1260 | 94 | |
XM_017006852.1 | XP_016862341.1 | 16 - 17 | 1678 | 94 | |
XM_017006853.1 | XP_016862342.1 | 3 - 4 | 367 | 94 | |
XM_017006854.1 | XP_016862343.1 | 3 - 4 | 370 | 94 | |
GenBank mRNA | AF516696.1 | - | 16 - 17 | 1606 | 94 |
AJ272213.1 | - | 14 - 15 | 1470 | 94 | |
AJ272268.1 | - | 15 - 16 | 1488 | 94 | |
AK299718.1 | - | 15 - 16 | 1470 | 94 | |
AK307449.1 | - | 14 - 15 | 1328 | 94 | |
BC137502.1 | - | 17 - 18 | 1676 | 94 | |
BC137505.1 | - | 17 - 18 | 1676 | 94 |