The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018398.2 | NP_060868.2 | 19 - 20 | 1828 | 107 |
XM_005265318.2 | XP_005265375.1 | 18 - 19 | 1882 | 107 | |
XM_011533946.2 | XP_011532248.1 | 19 - 20 | 1900 | 107 | |
XM_011533947.2 | XP_011532249.1 | 19 - 20 | 1900 | 107 | |
XM_011533948.2 | XP_011532250.1 | 19 - 20 | 1900 | 107 | |
XM_011533949.2 | XP_011532251.1 | 19 - 20 | 1900 | 107 | |
XM_011533954.2 | XP_011532256.1 | 6 - 7 | 1121 | 107 | |
XM_011533955.1 | XP_011532257.1 | 4 - 5 | 486 | 107 | |
XM_017006850.1 | XP_016862339.1 | 17 - 18 | 1810 | 107 | |
XM_017006851.1 | XP_016862340.1 | 16 - 17 | 1482 | 107 | |
XM_017006852.1 | XP_016862341.1 | 19 - 20 | 1900 | 107 | |
XM_017006853.1 | XP_016862342.1 | 6 - 7 | 589 | 107 | |
XM_017006854.1 | XP_016862343.1 | 6 - 7 | 592 | 107 | |
GenBank mRNA | AF516696.1 | - | 19 - 20 | 1828 | 107 |
AJ272268.1 | - | 18 - 19 | 1710 | 107 | |
AK307449.1 | - | 17 - 18 | 1550 | 107 | |
BC137502.1 | - | 20 - 21 | 1898 | 107 | |
BC137505.1 | - | 20 - 21 | 1898 | 107 |