The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018398.2 | NP_060868.2 | 28 - 29 | 2559 | 100 |
XM_005265318.2 | XP_005265375.1 | 27 - 28 | 2613 | 100 | |
XM_011533946.2 | XP_011532248.1 | 26 - 27 | 2487 | 100 | |
XM_011533947.2 | XP_011532249.1 | 28 - 29 | 2631 | 100 | |
XM_011533954.2 | XP_011532256.1 | 15 - 16 | 1852 | 100 | |
XM_011533955.1 | XP_011532257.1 | 13 - 14 | 1217 | 100 | |
XM_017006850.1 | XP_016862339.1 | 26 - 27 | 2541 | 100 | |
XM_017006851.1 | XP_016862340.1 | 25 - 26 | 2213 | 100 | |
XM_017006853.1 | XP_016862342.1 | 15 - 16 | 1320 | 100 | |
XM_017006854.1 | XP_016862343.1 | 15 - 16 | 1323 | 100 | |
GenBank mRNA | AF516696.1 | - | 28 - 29 | 2559 | 100 |
AJ272213.1 | - | 25 - 26 | 2361 | 100 | |
AJ272268.1 | - | 27 - 28 | 2441 | 100 | |
BC137502.1 | - | 29 - 30 | 2629 | 100 | |
BC137505.1 | - | 29 - 30 | 2629 | 100 | |
BI753834.1 | - | 2 - 3 | 111 | 100 |