The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018398.2 | NP_060868.2 | 31 - 32 | 2729 | 75 |
XM_005265318.2 | XP_005265375.1 | 30 - 31 | 2783 | 75 | |
XM_011533946.2 | XP_011532248.1 | 29 - 30 | 2657 | 75 | |
XM_011533947.2 | XP_011532249.1 | 31 - 32 | 2801 | 75 | |
XM_011533954.2 | XP_011532256.1 | 18 - 19 | 2022 | 75 | |
XM_011533955.1 | XP_011532257.1 | 16 - 17 | 1387 | 75 | |
XM_017006850.1 | XP_016862339.1 | 29 - 30 | 2711 | 75 | |
XM_017006851.1 | XP_016862340.1 | 28 - 29 | 2383 | 75 | |
XM_017006853.1 | XP_016862342.1 | 18 - 19 | 1490 | 75 | |
XM_017006854.1 | XP_016862343.1 | 18 - 19 | 1493 | 75 | |
GenBank mRNA | AF516696.1 | - | 31 - 32 | 2729 | 75 |
AJ272213.1 | - | 28 - 29 | 2531 | 75 | |
AJ272268.1 | - | 30 - 31 | 2611 | 75 | |
BC137502.1 | - | 32 - 33 | 2799 | 75 | |
BC137505.1 | - | 32 - 33 | 2799 | 75 | |
BG213431.1 | - | 2 - 3 | 75 | ||
BI753834.1 | - | 5 - 6 | 281 | 75 |