Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020410.2 NP_065143.2 12 - 13 1687 92
GenBank mRNA AB058728.1 - 12 - 13 1657 92
AF288687.1 - 12 - 13 1305 92
AK056420.1 - 12 - 13 1663 92
AK074159.1 - 5 - 6 723 92
AK074798.1 - 1 - 2 122 92
AK074920.1 - 1 - 2 337 92
AK130125.1 - 3 - 4 327 92
AK172778.1 - 11 - 12 1118 92

View Full Product Details