Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020410.2 NP_065143.2 20 - 21 2828 111
GenBank mRNA AB058728.1 - 20 - 21 2798 111
AF288687.1 - 20 - 21 2446 111
AK026044.1 - 4 - 5 978 111
AK056420.1 - 20 - 21 2804 111
AK095287.1 - 3 - 4 2001 111
AK123780.1 - 3 - 4 1172 111
AK125861.1 - 4 - 5 3125 111
AK130125.1 - 11 - 12 1468 111
AK172778.1 - 19 - 20 2259 111
BC009302.2 - 2 - 3 272 111
BC069211.1 - 1 - 2 162 111

View Full Product Details