Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
Chromosome Location
Chr.19: 19645198 - 19663694 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020410.2 NP_065143.2 23 - 24 3282 85
GenBank mRNA AB058728.1 - 23 - 24 3252 85
AF288687.1 - 23 - 24 2900 85
AK026044.1 - 7 - 8 1432 85
AK056420.1 - 23 - 24 3258 85
AK074159.1 - 14 - 15 4067 85
AK074798.1 - 11 - 12 1787 85
AK074920.1 - 12 - 13 1862 85
AK095287.1 - 6 - 7 2455 85
AK123780.1 - 6 - 7 1626 85
AK125861.1 - 7 - 8 3579 85
AK130125.1 - 14 - 15 1922 85
AK172778.1 - 22 - 23 2713 85
BC009302.2 - 5 - 6 726 85
BC069211.1 - 4 - 5 616 85

View Full Product Details