The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020410.2 | NP_065143.2 | 8 - 9 | 1241 | 110 |
GenBank mRNA | AB058728.1 | - | 8 - 9 | 1211 | 110 |
AF288687.1 | - | 8 - 9 | 859 | 110 | |
AK056420.1 | - | 8 - 9 | 1217 | 110 | |
AK074159.1 | - | 2 - 3 | 203 | 110 | |
AK172778.1 | - | 7 - 8 | 672 | 110 |