Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
Chromosome Location
Chr.14: 21016763 - 21070872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282211.1 NP_001269140.1 11 - 12 816 104
NM_001282213.1 NP_001269142.1 12 - 13 1095 104
NM_001282214.1 NP_001269143.1 12 - 13 1083 104
NM_001282216.1 NP_001269145.1 11 - 12 831 104
NM_001320329.1 NP_001307258.1 12 - 13 1748 104
NM_016250.2 NP_057334.1 12 - 13 938 104
NM_201535.1 NP_963293.1 13 - 14 980 104
NM_201536.1 NP_963294.1 11 - 12 871 104
NM_201537.1 NP_963831.1 12 - 13 913 104
NM_201538.1 NP_963832.1 11 - 12 848 104
NM_201539.1 NP_963833.1 12 - 13 890 104
NM_201540.1 NP_963834.1 13 - 14 957 104
NM_201541.1 NP_963835.1 12 - 13 915 104
XM_011536996.2 XP_011535298.1 13 - 14 967 104
XM_011536997.1 XP_011535299.1 13 - 14 968 104
XM_011536998.1 XP_011535300.1 13 - 14 958 104
XM_011536999.1 XP_011535301.1 11 - 12 1363 104
XM_011537001.1 XP_011535303.1 12 - 13 921 104
XM_011537002.1 XP_011535304.1 12 - 13 1067 104
GenBank mRNA AB033074.1 - 11 - 12 800 104
AF087872.1 - 11 - 12 854 104
AF159092.3 - 12 - 13 861 104
AK024521.1 - 13 - 14 913 104
AK057843.1 - 12 - 13 884 104
AK057847.1 - 11 - 12 848 104
AK090657.1 - 11 - 12 787 104
AK096999.1 - 12 - 13 1748 104
AK098388.1 - - 908 104
AK130029.1 - 11 - 12 786 104
AK293514.1 - 11 - 12 816 104
AL136574.1 - 11 - 12 825 104
BC010458.1 - 11 - 12 819 104
BC093038.1 - 12 - 13 914 104
BX248031.1 - 9 - 10 690 104
BX647091.1 - 9 - 10 1013 104
BX647748.1 - 12 - 13 1030 104
CR749252.1 - 12 - 13 1083 104

View Full Product Details