Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
Chromosome Location
Chr.14: 21016763 - 21070872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282211.1 NP_001269140.1 4 - 5 354 76
NM_001282212.1 NP_001269141.1 5 - 6 628 76
NM_001282213.1 NP_001269142.1 5 - 6 633 76
NM_001282214.1 NP_001269143.1 5 - 6 621 76
NM_001282215.1 NP_001269144.1 5 - 6 451 76
NM_001282216.1 NP_001269145.1 4 - 5 369 76
NM_001320329.1 NP_001307258.1 5 - 6 1286 76
NM_016250.2 NP_057334.1 5 - 6 476 76
NM_201535.1 NP_963293.1 6 - 7 518 76
NM_201536.1 NP_963294.1 4 - 5 409 76
NM_201537.1 NP_963831.1 5 - 6 451 76
NM_201538.1 NP_963832.1 4 - 5 386 76
NM_201539.1 NP_963833.1 5 - 6 428 76
NM_201540.1 NP_963834.1 6 - 7 495 76
NM_201541.1 NP_963835.1 5 - 6 453 76
XM_011536996.2 XP_011535298.1 6 - 7 505 76
XM_011536997.1 XP_011535299.1 6 - 7 506 76
XM_011536998.1 XP_011535300.1 6 - 7 496 76
XM_011536999.1 XP_011535301.1 4 - 5 901 76
XM_011537001.1 XP_011535303.1 5 - 6 459 76
XM_011537002.1 XP_011535304.1 5 - 6 605 76
XM_017021480.1 XP_016876969.1 6 - 7 457 76
XM_017021481.1 XP_016876970.1 6 - 7 496 76
XM_017021482.1 XP_016876971.1 6 - 7 505 76
XM_017021483.1 XP_016876972.1 5 - 6 389 76
XM_017021484.1 XP_016876973.1 5 - 6 431 76
XM_017021485.1 XP_016876974.1 6 - 7 496 76
XM_017021486.1 XP_016876975.1 6 - 7 506 76
XM_017021487.1 XP_016876976.1 5 - 6 415 76
XM_017021488.1 XP_016876977.1 5 - 6 456 76
XM_017021489.1 XP_016876978.1 5 - 6 561 76
XM_017021490.1 XP_016876979.1 4 - 5 353 76
XM_017021491.1 XP_016876980.1 4 - 5 388 76
XM_017021492.1 XP_016876981.1 5 - 6 459 76
XM_017021493.1 XP_016876982.1 5 - 6 605 76
XM_017021494.1 XP_016876983.1 6 - 7 457 76
XM_017021495.1 XP_016876984.1 5 - 6 389 76
XM_017021496.1 XP_016876985.1 4 - 5 388 76
GenBank mRNA AB033074.1 - 4 - 5 338 76
AF087872.1 - 4 - 5 392 76
AF159092.3 - 5 - 6 399 76
AK024521.1 - 6 - 7 451 76
AK057843.1 - 5 - 6 422 76
AK057847.1 - 4 - 5 386 76
AK090657.1 - 4 - 5 325 76
AK096999.1 - 5 - 6 1286 76
AK130029.1 - 4 - 5 324 76
AK293514.1 - 4 - 5 354 76
AL136574.1 - 4 - 5 363 76
AW163815.1 - 5 - 6 397 76
BC010458.1 - 4 - 5 357 76
BC011240.1 - 5 - 6 411 76
BG818977.1 - 5 - 6 432 76
BI759006.1 - 6 - 7 437 76
BM810623.1 - 5 - 6 392 76
BX247987.1 - 5 - 6 487 76
BX248031.1 - 3 - 4 315 76
BX647091.1 - 2 - 3 551 76
BX647748.1 - 5 - 6 568 76
CB153966.1 - 4 - 5 452 76
CR749252.1 - 5 - 6 621 76

View Full Product Details