Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NDRG2
- Entrez Gene ID
-
57447
- Gene Name
- NDRG family member 2
- Gene Aliases
-
SYLD
- Chromosome Location
-
Chr.14: 21016763 - 21070872 on Build GRCh38
View Full Product Details