Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
Chromosome Location
Chr.14: 21016763 - 21070872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282211.1 NP_001269140.1 9 - 10 729 80
NM_001282212.1 NP_001269141.1 10 - 11 1003 80
NM_001282213.1 NP_001269142.1 10 - 11 1008 80
NM_001282214.1 NP_001269143.1 10 - 11 996 80
NM_001282215.1 NP_001269144.1 10 - 11 826 80
NM_001282216.1 NP_001269145.1 9 - 10 744 80
NM_001320329.1 NP_001307258.1 10 - 11 1661 80
NM_016250.2 NP_057334.1 10 - 11 851 80
NM_201535.1 NP_963293.1 11 - 12 893 80
NM_201536.1 NP_963294.1 9 - 10 784 80
NM_201537.1 NP_963831.1 10 - 11 826 80
NM_201538.1 NP_963832.1 9 - 10 761 80
NM_201539.1 NP_963833.1 10 - 11 803 80
NM_201540.1 NP_963834.1 11 - 12 870 80
NM_201541.1 NP_963835.1 10 - 11 828 80
XM_011536996.2 XP_011535298.1 11 - 12 880 80
XM_011536997.1 XP_011535299.1 11 - 12 881 80
XM_011536998.1 XP_011535300.1 11 - 12 871 80
XM_011536999.1 XP_011535301.1 9 - 10 1276 80
XM_011537001.1 XP_011535303.1 10 - 11 834 80
XM_011537002.1 XP_011535304.1 10 - 11 980 80
XM_017021480.1 XP_016876969.1 11 - 12 832 80
XM_017021481.1 XP_016876970.1 11 - 12 871 80
XM_017021482.1 XP_016876971.1 11 - 12 880 80
XM_017021483.1 XP_016876972.1 10 - 11 764 80
XM_017021484.1 XP_016876973.1 10 - 11 806 80
XM_017021485.1 XP_016876974.1 11 - 12 871 80
XM_017021486.1 XP_016876975.1 11 - 12 881 80
XM_017021487.1 XP_016876976.1 10 - 11 790 80
XM_017021488.1 XP_016876977.1 10 - 11 831 80
XM_017021489.1 XP_016876978.1 10 - 11 936 80
XM_017021490.1 XP_016876979.1 9 - 10 728 80
XM_017021491.1 XP_016876980.1 9 - 10 763 80
XM_017021492.1 XP_016876981.1 10 - 11 834 80
XM_017021493.1 XP_016876982.1 10 - 11 980 80
XM_017021494.1 XP_016876983.1 11 - 12 832 80
XM_017021495.1 XP_016876984.1 10 - 11 764 80
XM_017021496.1 XP_016876985.1 9 - 10 763 80
XM_017021497.1 XP_016876986.1 5 - 6 361 80
GenBank mRNA AB033074.1 - 9 - 10 713 80
AF087872.1 - 9 - 10 767 80
AF159092.3 - 10 - 11 774 80
AK024521.1 - 11 - 12 826 80
AK057843.1 - 10 - 11 797 80
AK057847.1 - 9 - 10 761 80
AK090657.1 - 9 - 10 700 80
AK096999.1 - 10 - 11 1661 80
AK098388.1 - - 821 80
AK130029.1 - 9 - 10 699 80
AK293514.1 - 9 - 10 729 80
AL136574.1 - 9 - 10 738 80
BC010458.1 - 9 - 10 732 80
BC011240.1 - 10 - 11 786 80
BC013209.1 - 2 - 3 857 80
BC093038.1 - 10 - 11 827 80
BM810623.1 - 10 - 11 767 80
BX247987.1 - 10 - 11 862 80
BX248031.1 - 7 - 8 603 80
BX647091.1 - 7 - 8 926 80
BX647748.1 - 10 - 11 943 80
CR749252.1 - 10 - 11 996 80

View Full Product Details