Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM81B
Entrez Gene ID
153643
Gene Name
family with sequence similarity 81 member B
Gene Aliases
-
Chromosome Location
Chr.5: 95384793 - 95450441 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152548.2 NP_689761.2 8 - 9 1078 96
XM_005271905.2 XP_005271962.2 7 - 8 937 96
XM_005271906.4 XP_005271963.2 10 - 11 1242 96
XM_011543209.1 XP_011541511.1 6 - 7 759 96
XM_011543210.2 XP_011541512.1 5 - 6 792 96
XM_017009118.1 XP_016864607.1 9 - 10 1098 96
GenBank mRNA AK058062.1 - 8 - 9 1061 96
AK310245.1 - 8 - 9 1021 96
BC034772.1 - 8 - 9 1078 96

View Full Product Details