The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293306.2 | NP_001280235.2 | 12 - 13 | 1862 | 72 |
NM_001293307.2 | NP_001280236.2 | 11 - 12 | 1568 | 72 | |
NM_006514.3 | NP_006505.3 | 12 - 13 | 1862 | 72 | |
XM_005265371.3 | XP_005265428.1 | 12 - 13 | 1871 | 72 | |
XM_011533993.2 | XP_011532295.1 | 12 - 13 | 1871 | 72 | |
XM_011533994.2 | XP_011532296.1 | 11 - 12 | 1577 | 72 | |
GenBank mRNA | AF117907.1 | - | 12 - 13 | 1862 | 72 |