The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252385.1 | NP_001239314.1 | 1 - 2 | 208 | 83 |
NM_001252393.1 | NP_001239322.1 | 1 - 2 | 208 | 83 | |
NM_006058.4 | NP_006049.3 | 1 - 2 | 208 | 83 | |
XM_005268355.1 | XP_005268412.1 | 1 - 2 | 121 | 83 | |
XM_006714751.1 | XP_006714814.1 | 1 - 2 | 121 | 83 | |
XM_006714752.2 | XP_006714815.1 | 1 - 2 | 121 | 83 | |
XM_017008945.1 | XP_016864434.1 | 1 - 2 | 121 | 83 | |
XM_017008949.1 | XP_016864438.1 | 1 - 2 | 121 | 83 | |
XM_017008950.1 | XP_016864439.1 | 1 - 2 | 121 | 83 | |
GenBank mRNA | AB252972.1 | - | 1 - 2 | 198 | 83 |
AB252973.1 | - | 1 - 2 | 198 | 83 | |
AK074400.1 | - | 1 - 2 | 139 | 83 | |
AK303573.1 | - | 1 - 2 | 158 | 83 | |
BC012133.1 | - | 1 - 2 | 114 | 83 | |
BP254790.1 | - | 1 - 2 | 198 | 83 | |
DC374977.1 | - | 1 - 2 | 208 | 83 |