Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPO1
Entrez Gene ID
284654
Gene Name
R-spondin 1
Gene Aliases
CRISTIN3, RSPO
Chromosome Location
Chr.1: 37611350 - 37634923 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038633.3 NP_001033722.1 7 - 8 1512 58
NM_001242908.1 NP_001229837.1 6 - 7 1333 58
NM_001242909.1 NP_001229838.1 6 - 7 1012 58
XM_006710583.3 XP_006710646.1 5 - 6 1248 58
GenBank mRNA AK098225.1 - 6 - 7 754 58
BC114966.1 - 5 - 6 930 58
DQ165084.1 - 6 - 7 1075 58
DQ318235.1 - 7 - 8 1222 58

View Full Product Details