Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC38A4
Entrez Gene ID
55089
Gene Name
solute carrier family 38 member 4
Gene Aliases
ATA3, NAT3, PAAT, SNAT4
Chromosome Location
Chr.12: 46764761 - 46832422 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143824.1 NP_001137296.1 7 - 8 779 117
NM_018018.4 NP_060488.2 8 - 9 971 117
XM_005268997.2 XP_005269054.1 7 - 8 780 117
GenBank mRNA AB055003.1 - 6 - 7 603 117
AF193836.1 - 7 - 8 759 117
AF305814.1 - 8 - 9 936 117
AK001053.1 - 8 - 9 847 117
AK291168.1 - 7 - 8 780 117
BC069819.1 - 7 - 8 832 117
BC101827.1 - 6 - 7 616 117
BC104913.1 - 6 - 7 615 117

View Full Product Details