The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077242.1 | NP_001070710.1 | 2 - 3 | 555 | 87 |
NM_139160.2 | NP_631899.2 | 2 - 3 | 794 | 87 | |
GenBank mRNA | AF447588.1 | - | 2 - 3 | 794 | 87 |
AJ245600.1 | - | 2 - 3 | 510 | 87 | |
AK295960.1 | - | 2 - 3 | 508 | 87 | |
BC030970.1 | - | 2 - 3 | 555 | 87 |