The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012210.3 | NP_036342.2 | 1 - 2 | 74 | 64 |
GenBank mRNA | AK314264.1 | - | 1 - 2 | 55 | 64 |
DN991209.1 | - | 1 - 2 | 36 | 64 |