Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDAC7
Entrez Gene ID
51564
Gene Name
histone deacetylase 7
Gene Aliases
HD7, HD7A, HDAC7A
Chromosome Location
Chr.12: 47782710 - 47820612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098416.3 NP_001091886.1 11 - 12 1547 121
NM_001308090.1 NP_001295019.1 11 - 12 1607 121
NM_015401.4 NP_056216.2 12 - 13 1658 121
XM_011538478.1 XP_011536780.1 12 - 13 1610 121
XM_011538479.1 XP_011536781.1 13 - 14 2052 121
XM_011538480.1 XP_011536782.1 11 - 12 1499 121
XM_011538481.1 XP_011536783.1 12 - 13 1503 121
XM_011538482.1 XP_011536784.1 12 - 13 1587 121
XM_011538483.1 XP_011536785.1 12 - 13 1610 121
XM_017019455.1 XP_016874944.1 11 - 12 1559 121
XM_017019456.1 XP_016874945.1 12 - 13 1610 121
GenBank mRNA AF239243.1 - 9 - 10 1441 121
AK001190.1 - 4 - 5 580 121
AK024469.1 - 7 - 8 973 121
AK299292.1 - 12 - 13 1573 121
AK301545.1 - 12 - 13 1521 121
AK303481.1 - 11 - 12 1513 121
AL117455.1 - 8 - 9 1125 121
AY302468.1 - 12 - 13 1463 121
AY321367.1 - 11 - 12 1432 121
BC020505.2 - 4 - 5 625 121
BC064840.1 - 11 - 12 1441 121
BT009771.1 - 2 - 3 332 121

View Full Product Details