The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002442.3 | NP_002433.1 | 12 - 13 | 943 | 108 |
XM_006719404.3 | XP_006719467.1 | 7 - 8 | 722 | 108 | |
XM_011538361.2 | XP_011536663.1 | 12 - 13 | 937 | 108 | |
XM_011538362.2 | XP_011536664.1 | 11 - 12 | 880 | 108 | |
XM_011538364.2 | XP_011536666.1 | 8 - 9 | 1487 | 108 | |
XM_011538365.2 | XP_011536667.1 | 9 - 10 | 606 | 108 | |
XM_011538366.2 | XP_011536668.1 | 9 - 10 | 1614 | 108 | |
XM_011538368.2 | XP_011536670.1 | 7 - 8 | 562 | 108 | |
XM_011538370.2 | XP_011536672.1 | 7 - 8 | 488 | 108 | |
XM_011538371.2 | XP_011536673.1 | 7 - 8 | 514 | 108 | |
GenBank mRNA | AB012851.1 | - | 12 - 13 | 918 | 108 |
AK023262.1 | - | 5 - 6 | 363 | 108 |