Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELP4
Entrez Gene ID
26610
Gene Name
elongator acetyltransferase complex subunit 4
Gene Aliases
AN, C11orf19, PAX6NEB, PAXNEB, dJ68P15A.1, hELP4
Chromosome Location
Chr.11: 31509729 - 31784525 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288726.1 NP_001275655.1 4 - 5 566 110
NM_019040.4 NP_061913.3 4 - 5 566 110
GenBank mRNA AK000505.1 - 4 - 5 545 110
AL136677.1 - 4 - 5 523 110
BC012514.1 - 4 - 5 517 110

View Full Product Details