The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004626.2 | NP_004617.2 | 3 - 4 | 722 | 70 |
XM_005274231.1 | XP_005274288.1 | 4 - 5 | 978 | 70 | |
XM_011545238.1 | XP_011543540.1 | 4 - 5 | 1008 | 70 | |
XM_011545239.2 | XP_011543541.1 | 4 - 5 | 1216 | 70 | |
XM_011545240.2 | XP_011543542.1 | 5 - 6 | 1172 | 70 | |
XM_011545241.2 | XP_011543543.1 | 3 - 4 | 460 | 70 | |
XM_017018241.1 | XP_016873730.1 | 2 - 3 | 1480 | 70 | |
GenBank mRNA | AB070218.1 | - | 3 - 4 | 661 | 70 |
AK313570.1 | - | 3 - 4 | 724 | 70 | |
BC074790.2 | - | 3 - 4 | 641 | 70 | |
BC074791.2 | - | 3 - 4 | 641 | 70 | |
BC113386.1 | - | 3 - 4 | 642 | 70 | |
BC113388.1 | - | 3 - 4 | 642 | 70 | |
Y12692.1 | - | 3 - 4 | 722 | 70 |