Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VLDLR
Entrez Gene ID
7436
Gene Name
very low density lipoprotein receptor
Gene Aliases
CAMRQ1, CARMQ1, CHRMQ1, VLDL-R, VLDLRCH
Chromosome Location
Chr.9: 2621679 - 2656103 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001018056.2 NP_001018066.1 6 - 7 1359 75
NM_001322225.1 NP_001309154.1 5 - 6 1236 75
NM_001322226.1 NP_001309155.1 5 - 6 1236 75
NM_003383.4 NP_003374.3 6 - 7 1359 75
GenBank mRNA AB208822.1 - 7 - 8 1115 75
AK092381.1 - 4 - 5 626 75
AK095744.1 - 2 - 3 439 75
BC136562.1 - 6 - 7 1080 75
BC144245.1 - 6 - 7 1080 75
D16493.1 - 6 - 7 1013 75
D16494.1 - 6 - 7 944 75
L20470.1 - 6 - 7 1334 75
L22431.1 - 6 - 7 1056 75
S73849.1 - 6 - 7 1028 75

View Full Product Details