The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005651.3 | NP_005642.1 | 5 - 6 | 499 | 68 |
GenBank mRNA | AK289418.1 | - | 5 - 6 | 496 | 68 |
AK307741.1 | - | 5 - 6 | 496 | 68 | |
BC005355.1 | - | 5 - 6 | 496 | 68 | |
U32989.1 | - | 5 - 6 | 497 | 68 |