Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
Chromosome Location
Chr.17: 37084992 - 37406822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198834.2 NP_942131.1 30 - 31 4215 67
NM_198836.2 NP_942133.1 30 - 31 3783 67
NM_198837.1 NP_942134.1 28 - 29 3853 67
NM_198838.1 NP_942135.1 29 - 30 3964 67
NM_198839.2 NP_942136.1 34 - 35 4889 67
XM_005257267.4 XP_005257324.1 30 - 31 3696 67
XM_011524701.1 XP_011523003.1 30 - 31 4136 67
XM_011524703.1 XP_011523005.1 30 - 31 3768 67
XM_011524704.2 XP_011523006.1 30 - 31 3701 67
XM_017024553.1 XP_016880042.1 31 - 32 4250 67
XM_017024555.1 XP_016880044.1 30 - 31 3849 67
GenBank mRNA AB371587.1 - 30 - 31 3762 67
AY237919.1 - 28 - 29 3592 67
AY315627.1 - 28 - 29 3592 67
BC137287.1 - 30 - 31 3720 67
U19822.1 - 28 - 29 3592 67

View Full Product Details