Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
Chromosome Location
Chr.17: 37084992 - 37406822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198834.2 NP_942131.1 42 - 43 5856 99
NM_198836.2 NP_942133.1 42 - 43 5424 99
NM_198837.1 NP_942134.1 40 - 41 5494 99
NM_198838.1 NP_942135.1 41 - 42 5605 99
NM_198839.2 NP_942136.1 46 - 47 6530 99
XM_005257267.4 XP_005257324.1 42 - 43 5337 99
XM_006721853.1 XP_006721916.1 41 - 42 5843 99
XM_011524701.1 XP_011523003.1 42 - 43 5777 99
XM_011524703.1 XP_011523005.1 42 - 43 5409 99
XM_011524704.2 XP_011523006.1 42 - 43 5342 99
XM_017024553.1 XP_016880042.1 43 - 44 5891 99
XM_017024554.1 XP_016880043.1 42 - 43 5867 99
XM_017024555.1 XP_016880044.1 42 - 43 5490 99
GenBank mRNA AB209325.1 - 9 - 10 1189 99
AB371587.1 - 42 - 43 5403 99
AK295735.1 - 1 - 2 50 99
AY237919.1 - 40 - 41 5233 99
AY315627.1 - 40 - 41 5233 99
BC031485.1 - 1 - 2 266 99
BC137287.1 - 42 - 43 5361 99

View Full Product Details