Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
Chromosome Location
Chr.17: 37084992 - 37406822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198834.2 NP_942131.1 46 - 47 6336 101
NM_198836.2 NP_942133.1 46 - 47 5904 101
NM_198837.1 NP_942134.1 44 - 45 5974 101
NM_198838.1 NP_942135.1 45 - 46 6085 101
NM_198839.2 NP_942136.1 50 - 51 7010 101
XM_005257267.4 XP_005257324.1 46 - 47 5817 101
XM_006721853.1 XP_006721916.1 45 - 46 6323 101
XM_011524701.1 XP_011523003.1 46 - 47 6257 101
XM_011524703.1 XP_011523005.1 46 - 47 5889 101
XM_011524704.2 XP_011523006.1 46 - 47 5822 101
XM_017024553.1 XP_016880042.1 47 - 48 6371 101
XM_017024554.1 XP_016880043.1 46 - 47 6347 101
XM_017024555.1 XP_016880044.1 46 - 47 5970 101
GenBank mRNA AB209325.1 - 13 - 14 1669 101
AB371587.1 - 46 - 47 5883 101
AK295735.1 - 4 - 5 409 101
AY237919.1 - 44 - 45 5713 101
AY315627.1 - 44 - 45 5713 101
BC031485.1 - 5 - 6 746 101
BC137287.1 - 46 - 47 5841 101

View Full Product Details