Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
Chromosome Location
Chr.17: 37084992 - 37406822 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198834.2 NP_942131.1 48 - 49 6554 66
NM_198836.2 NP_942133.1 48 - 49 6122 66
NM_198837.1 NP_942134.1 46 - 47 6192 66
NM_198838.1 NP_942135.1 47 - 48 6303 66
NM_198839.2 NP_942136.1 52 - 53 7228 66
XM_005257267.4 XP_005257324.1 48 - 49 6035 66
XM_006721853.1 XP_006721916.1 47 - 48 6541 66
XM_011524701.1 XP_011523003.1 48 - 49 6475 66
XM_011524703.1 XP_011523005.1 48 - 49 6107 66
XM_011524704.2 XP_011523006.1 48 - 49 6040 66
XM_017024553.1 XP_016880042.1 49 - 50 6589 66
XM_017024554.1 XP_016880043.1 48 - 49 6565 66
XM_017024555.1 XP_016880044.1 48 - 49 6188 66
GenBank mRNA AB209325.1 - 15 - 16 1887 66
AB371587.1 - 48 - 49 6101 66
AK295735.1 - 6 - 7 627 66
AK295805.1 - 2 - 3 127 66
AK301579.1 - 2 - 3 228 66
AY237919.1 - 46 - 47 5931 66
AY315627.1 - 46 - 47 5931 66
BC031485.1 - 7 - 8 964 66
BC137287.1 - 48 - 49 6059 66

View Full Product Details