The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198834.2 | NP_942131.1 | 41 - 42 | 5589 | 88 |
NM_198836.2 | NP_942133.1 | 41 - 42 | 5157 | 88 | |
NM_198837.1 | NP_942134.1 | 39 - 40 | 5227 | 88 | |
NM_198838.1 | NP_942135.1 | 40 - 41 | 5338 | 88 | |
NM_198839.2 | NP_942136.1 | 45 - 46 | 6263 | 88 | |
XM_005257267.4 | XP_005257324.1 | 41 - 42 | 5070 | 88 | |
XM_006721853.1 | XP_006721916.1 | 40 - 41 | 5576 | 88 | |
XM_011524701.1 | XP_011523003.1 | 41 - 42 | 5510 | 88 | |
XM_011524703.1 | XP_011523005.1 | 41 - 42 | 5142 | 88 | |
XM_011524704.2 | XP_011523006.1 | 41 - 42 | 5075 | 88 | |
XM_017024553.1 | XP_016880042.1 | 42 - 43 | 5624 | 88 | |
XM_017024554.1 | XP_016880043.1 | 41 - 42 | 5600 | 88 | |
XM_017024555.1 | XP_016880044.1 | 41 - 42 | 5223 | 88 | |
GenBank mRNA | AB371587.1 | - | 41 - 42 | 5136 | 88 |
AY237919.1 | - | 39 - 40 | 4966 | 88 | |
AY315627.1 | - | 39 - 40 | 4966 | 88 | |
BC137287.1 | - | 41 - 42 | 5094 | 88 | |
U19822.1 | - | 39 - 40 | 4966 | 88 |