The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040167.1 | NP_001035257.1 | 6 - 7 | 1003 | 109 |
NM_001040168.1 | NP_001035258.1 | 6 - 7 | 1003 | 109 | |
NM_001166355.1 | NP_001159827.1 | 7 - 8 | 899 | 109 | |
NM_002304.2 | NP_002295.1 | 7 - 8 | 687 | 109 | |
GenBank mRNA | AF193612.1 | - | 6 - 7 | 678 | 109 |
AK096284.1 | - | 7 - 8 | 899 | 109 | |
BC014851.1 | - | 7 - 8 | 687 | 109 | |
U94354.1 | - | 6 - 7 | 676 | 109 |