The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077628.2 | NP_001071096.1 | 2 - 3 | 684 | 67 |
NM_016022.3 | NP_057106.2 | 2 - 3 | 684 | 67 | |
NR_045033.1 | 2 - 3 | 548 | 67 | ||
GenBank mRNA | AF151835.1 | - | 2 - 3 | 386 | 67 |
AF508787.1 | - | 2 - 3 | 342 | 67 | |
AK027879.1 | - | 2 - 3 | 454 | 67 | |
AK075295.1 | - | 2 - 3 | 403 | 67 | |
AK075452.1 | - | 2 - 3 | 492 | 67 | |
AK297680.1 | - | 2 - 3 | 314 | 67 | |
AK314490.1 | - | 2 - 3 | 454 | 67 | |
AX775955.1 | - | 2 - 3 | 420 | 67 | |
AX775957.1 | - | 2 - 3 | 474 | 67 | |
AY113698.1 | - | 2 - 3 | 342 | 67 | |
AY113699.1 | - | 2 - 3 | 342 | 67 | |
BC001230.2 | - | 2 - 3 | 454 | 67 | |
BC008732.2 | - | 2 - 3 | 441 | 67 | |
BC009501.2 | - | 2 - 3 | 475 | 67 | |
BC015568.2 | - | 2 - 3 | 443 | 67 | |
BC017699.1 | - | 2 - 3 | 420 | 67 | |
BC020590.1 | - | 2 - 3 | 474 | 67 |