The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020693.3 | NP_065744.2 | 19 - 20 | 3751 | 146 |
XM_011542917.1 | XP_011541219.1 | 19 - 20 | 3781 | 146 | |
XM_011542918.1 | XP_011541220.1 | 20 - 21 | 3469 | 146 | |
XM_011542919.2 | XP_011541221.1 | 14 - 15 | 2424 | 146 | |
XM_011542920.2 | XP_011541222.1 | 14 - 15 | 2471 | 146 | |
XM_011542921.2 | XP_011541223.1 | 10 - 11 | 2028 | 146 | |
XM_011542924.2 | XP_011541226.1 | 9 - 10 | 1946 | 146 | |
XM_011542925.2 | XP_011541227.1 | 9 - 10 | 2620 | 146 | |
GenBank mRNA | AB032958.2 | - | 19 - 20 | 3686 | 146 |
AF304304.1 | - | 19 - 20 | 3581 | 146 | |
AF304305.1 | - | 17 - 18 | 2948 | 146 | |
AF334384.1 | - | 19 - 20 | 3663 | 146 | |
AF491813.1 | - | 19 - 20 | 3751 | 146 |