Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSCAML1
Entrez Gene ID
57453
Gene Name
DS cell adhesion molecule like 1
Gene Aliases
DSCAM2
Chromosome Location
Chr.11: 117427772 - 117797261 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020693.3 NP_065744.2 27 - 28 5059 86
XM_011542917.1 XP_011541219.1 27 - 28 5089 86
XM_011542918.1 XP_011541220.1 28 - 29 4777 86
XM_011542919.2 XP_011541221.1 22 - 23 3732 86
XM_011542920.2 XP_011541222.1 22 - 23 3779 86
XM_011542921.2 XP_011541223.1 18 - 19 3336 86
XM_011542924.2 XP_011541226.1 17 - 18 3254 86
XM_011542925.2 XP_011541227.1 17 - 18 3928 86
GenBank mRNA AB032958.2 - 27 - 28 4994 86
AF304304.1 - 27 - 28 4889 86
AF304305.1 - 25 - 26 4256 86
AF334384.1 - 27 - 28 4971 86
AF491813.1 - 27 - 28 5059 86

View Full Product Details