The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278585.1 | NP_001265514.1 | 14 - 15 | 2044 | 72 |
NM_006587.3 | NP_006578.2 | 16 - 17 | 2356 | 72 | |
GenBank mRNA | AF133845.1 | - | 16 - 17 | 2292 | 72 |
AK302816.1 | - | 15 - 16 | 2031 | 72 | |
AK304038.1 | - | 15 - 16 | 2121 | 72 | |
BC110451.2 | - | 16 - 17 | 2292 | 72 |