Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NT5DC2
Entrez Gene ID
64943
Gene Name
5'-nucleotidase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 52524369 - 52535077 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134231.1 NP_001127703.1 2 - 3 455 67
NM_022908.2 NP_075059.1 2 - 3 728 67
XM_005265406.2 XP_005265463.1 2 - 3 535 67
XM_006713303.3 XP_006713366.1 2 - 3 449 67
XM_006713305.2 XP_006713368.1 2 - 3 278 67
GenBank mRNA AK022504.1 - 2 - 3 258 67
AK023995.1 - 2 - 3 715 67
AW510639.1 - 2 - 3 394 67
DR003312.1 - 2 - 3 462 67

View Full Product Details