The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134231.1 | NP_001127703.1 | 7 - 8 | 868 | 67 |
NM_022908.2 | NP_075059.1 | 7 - 8 | 1141 | 67 | |
XM_005265406.2 | XP_005265463.1 | 7 - 8 | 948 | 67 | |
XM_006713303.3 | XP_006713366.1 | 7 - 8 | 862 | 67 | |
XM_006713305.2 | XP_006713368.1 | 7 - 8 | 691 | 67 | |
GenBank mRNA | AF131781.1 | - | 4 - 5 | 410 | 67 |
AK022504.1 | - | 7 - 8 | 671 | 67 | |
AK023995.1 | - | 7 - 8 | 1128 | 67 | |
BC014550.1 | - | 6 - 7 | 550 | 67 |