Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH3PXD2A
Entrez Gene ID
9644
Gene Name
SH3 and PX domains 2A
Gene Aliases
FISH, SH3MD1, TKS5
Chromosome Location
Chr.10: 103594027 - 103855464 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270294.4 XP_005270351.1 10 - 11 984 57
XM_005270295.4 XP_005270352.1 9 - 10 938 57
XM_005270297.3 XP_005270354.1 4 - 5 2215 57
XM_005270298.4 XP_005270355.1 4 - 5 1018 57
XM_006718079.3 XP_006718142.1 11 - 12 1081 57
XM_006718080.3 XP_006718143.1 5 - 6 1112 57
XM_011540385.2 XP_011538687.1 10 - 11 1035 57
XM_011540386.2 XP_011538688.1 5 - 6 2311 57
XM_017016970.1 XP_016872459.1 9 - 10 818 57
GenBank mRNA AK294454.1 - 4 - 5 391 57
AK295314.1 - 4 - 5 468 57
AK302015.1 - 7 - 8 607 57
AK304828.1 - 4 - 5 407 57

View Full Product Details