The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014631.2 | NP_055446.2 | 11 - 12 | 1317 | 66 |
XM_005270294.4 | XP_005270351.1 | 12 - 13 | 1450 | 66 | |
XM_005270295.4 | XP_005270352.1 | 11 - 12 | 1404 | 66 | |
XM_005270297.3 | XP_005270354.1 | 6 - 7 | 2681 | 66 | |
XM_005270298.4 | XP_005270355.1 | 6 - 7 | 1484 | 66 | |
XM_006718079.3 | XP_006718142.1 | 13 - 14 | 1547 | 66 | |
XM_006718080.3 | XP_006718143.1 | 7 - 8 | 1578 | 66 | |
XM_011540385.2 | XP_011538687.1 | 12 - 13 | 1501 | 66 | |
XM_011540386.2 | XP_011538688.1 | 7 - 8 | 2777 | 66 | |
XM_011540387.2 | XP_011538689.1 | 3 - 4 | 4423 | 66 | |
XM_017016969.1 | XP_016872458.1 | 12 - 13 | 1468 | 66 | |
XM_017016970.1 | XP_016872459.1 | 11 - 12 | 1284 | 66 | |
GenBank mRNA | AB007878.1 | - | 9 - 10 | 1067 | 66 |
AK056469.1 | - | 10 - 11 | 1040 | 66 | |
AK294454.1 | - | 6 - 7 | 857 | 66 | |
AK294736.1 | - | 6 - 7 | 895 | 66 | |
AK295314.1 | - | 6 - 7 | 934 | 66 | |
AK295660.1 | - | 6 - 7 | 961 | 66 | |
AK302015.1 | - | 9 - 10 | 1073 | 66 | |
AK304828.1 | - | 6 - 7 | 873 | 66 | |
AK315983.1 | - | 6 - 7 | 900 | 66 |