The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003939.4 | NP_003930.1 | 2 - 3 | 276 | 126 |
NM_033637.3 | NP_378663.1 | 3 - 4 | 384 | 126 | |
XM_006718054.2 | XP_006718117.1 | 3 - 4 | 684 | 126 | |
XM_017016871.1 | XP_016872360.1 | 3 - 4 | 275 | 126 | |
XM_017016872.1 | XP_016872361.1 | 3 - 4 | 447 | 126 | |
XM_017016873.1 | XP_016872362.1 | 3 - 4 | 303 | 126 | |
GenBank mRNA | AF101784.1 | - | 3 - 4 | 236 | 126 |
AF129530.1 | - | 2 - 3 | 197 | 126 | |
AK313353.1 | - | 3 - 4 | 257 | 126 | |
AK313417.1 | - | 2 - 3 | 240 | 126 | |
AL707686.1 | - | 3 - 4 | 353 | 126 | |
BC027994.1 | - | 3 - 4 | 273 | 126 | |
GU727631.1 | - | 3 - 4 | 273 | 126 | |
KU178451.1 | - | 3 - 4 | 236 | 126 | |
KU178452.1 | - | 5 - 6 | 385 | 126 | |
KU178453.1 | - | 4 - 5 | 329 | 126 | |
Y14153.1 | - | 2 - 3 | 197 | 126 |