The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256856.1 | NP_001243785.1 | 8 - 9 | 1169 | 114 |
NM_003939.4 | NP_003930.1 | 8 - 9 | 1139 | 114 | |
NM_033637.3 | NP_378663.1 | 9 - 10 | 1247 | 114 | |
XM_006718054.2 | XP_006718117.1 | 9 - 10 | 1547 | 114 | |
XM_011540320.2 | XP_011538622.1 | 6 - 7 | 983 | 114 | |
XM_017016870.1 | XP_016872359.1 | 8 - 9 | 1469 | 114 | |
XM_017016871.1 | XP_016872360.1 | 9 - 10 | 1138 | 114 | |
XM_017016872.1 | XP_016872361.1 | 9 - 10 | 1310 | 114 | |
XM_017016873.1 | XP_016872362.1 | 9 - 10 | 1166 | 114 | |
XM_017016874.1 | XP_016872363.1 | 7 - 8 | 1061 | 114 | |
GenBank mRNA | AF101784.1 | - | 9 - 10 | 1099 | 114 |
AF129530.1 | - | 8 - 9 | 1060 | 114 | |
AK295870.1 | - | 8 - 9 | 1037 | 114 | |
AK313353.1 | - | 9 - 10 | 1120 | 114 | |
AK313417.1 | - | 8 - 9 | 1103 | 114 | |
BC027994.1 | - | 9 - 10 | 1136 | 114 | |
CR749295.1 | - | 8 - 9 | 1100 | 114 | |
GU727631.1 | - | 9 - 10 | 1136 | 114 | |
KU178451.1 | - | 9 - 10 | 1099 | 114 | |
KU178452.1 | - | 11 - 12 | 1248 | 114 | |
KU178453.1 | - | 10 - 11 | 1192 | 114 | |
KU178454.1 | - | 8 - 9 | 1021 | 114 | |
Y14153.1 | - | 8 - 9 | 1060 | 114 |